Skip to Content
Merck
Ethyl 6-bromohexanoate 99%

Ethyl 6-bromohexanoate

Ethyl 6-bromohexanoate 99%
Synonym(s):
6-Bromohexanoic acid ethyl ester, Ethyl 6-bromocapronate
Linear Formula:
Br(CH2)5COOCH2CH3
CAS Number:
Molecular Weight:
223.11
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing