
Estradiol cypionate

Synonym(s):
β-Estradiol 17-cypionate, 1,3,5(10)-Estratriene-3,17β-diol 17-cyclopentylpropionate, 3,17β-Hydroxy-1,3,5(10)-estratriene 17-cyclopentylpropionate, Depoestradiol
Empirical Formula (Hill Notation):
C26H36O3
CAS Number:
Molecular Weight:
396.56
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing