Eppendorf® Deepwell plates, standard, 96 wells
Products
Product Category
Manufacturer
Wells
Working Volume (ml)
Color
Package Size
Sterility
Sterilization Method
Technique
Showing 1-20 of 29
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2