
Enalapril maleate

Synonym(s):
Enalapril maleate salt, (S)-N-[1-(Ethoxycarbonyl)-3-phenylpropyl]-Ala-Pro maleate salt
Empirical Formula (Hill Notation):
C20H28N2O5 · C4H4O4
CAS Number:
Molecular Weight:
492.52
Products
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing