
Docusate sodium

Synonym(s):
Docusate sodium salt, AOT, Bis(2-ethylhexyl) sulfosuccinate sodium salt, Sodium bis(2-ethylhexyl) sulfosuccinate, Sulfobutanedioic acid bis(2-ethylhexyl ester) sodium salt, Sulfosuccinic acid bis(2-ethylhexyl) ester sodium salt, ‘Dioctyl’ sulfosuccinate sodium salt
Empirical Formula (Hill Notation):
C20H37NaO7S
Products
Product Category
Feature
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing