
DMT-2′O-TBDMS-rU Phosphoramidite

Synonym(s):
5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-uridine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], DMT-2′-O-TBDMS-rU amidite
Empirical Formula (Hill Notation):
C45H61N4O9PSi
CAS Number:
Molecular Weight:
861.05
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique