(±)-L-Alliin
Synonym(s):
S-Allyl-L-cysteine sulfoxide
Empirical Formula (Hill Notation):
C6H11NO3S
CAS Number:
Molecular Weight:
177.22
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing