
Diphenylamine-4-sulfonic acid barium salt

Synonym(s):
Diphenylamine-4-sulfonic acid barium salt
Empirical Formula (Hill Notation):
C24H20BaN2O6S2
CAS Number:
Molecular Weight:
633.88
Products
Brand
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing