
Dimethyl fumarate

Synonym(s):
Dimethyl fumarate
Linear Formula:
CH3OCOCH=CHCOOCH3
CAS Number:
Molecular Weight:
144.13
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing