
Dimethyl 2,6-dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylate

Empirical Formula (Hill Notation):
C17H16N2O5
CAS Number:
Molecular Weight:
328.32
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing