
Diethanolamine fusidate

Empirical Formula (Hill Notation):
C35H59NO8
CAS Number:
Molecular Weight:
621.84
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing