
Dantrolene

Synonym(s):
1-{{[5-(4-Nitrophenyl)-2-furanyl]methylene}amino}-2,4-imidazolidinedione, Dantrolene
Empirical Formula (Hill Notation):
C14H10N4O5
CAS Number:
Molecular Weight:
314.25
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing