Cyclopropylmagnesium bromide solution
Empirical Formula (Hill Notation):
C3H5BrMg
CAS Number:
Molecular Weight:
145.28
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing