
Cyclobenzaprine hydrochloride

Synonym(s):
5-(3-Dimethylaminopropylidene)dibenzo[a,e]cycloheptatriene hydrochloride
Empirical Formula (Hill Notation):
C20H21N · HCl
CAS Number:
Molecular Weight:
311.85
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing