Chebulagic acid
Empirical Formula (Hill Notation):
C41H30O27
CAS Number:
Molecular Weight:
954.66
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing