
Cefotaxime for peak identification

Synonym(s):
Cefotaxime sodium salt, Cefotaxim sodium salt
Empirical Formula (Hill Notation):
C16H16N5NaO7S2
CAS Number:
Molecular Weight:
477.45
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing