Skip to Content
Merck
Cefazolin Pharmaceutical Secondary Standard; Certified Reference Material

Cefazolin

Cefazolin Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Cephazolin, Cefazolin
Empirical Formula (Hill Notation):
C14H14N8O4S3
CAS Number:
Molecular Weight:
454.51

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade