Skip to Content
Merck
Bromoxylenol Blue indicator grade, Dye content 95 %

Bromoxylenol Blue

Bromoxylenol Blue indicator grade, Dye content 95 %
Synonym(s):
3′,3″-Dibromo-p-xylenolsulfonphthalein
Empirical Formula (Hill Notation):
C23H20Br2O5S
CAS Number:
Molecular Weight:
568.27
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Chemical Composition
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing