
Boc-D-Ile-OH

Synonym(s):
Boc-D-isoleucine
Empirical Formula (Hill Notation):
C11H21NO4
CAS Number:
Molecular Weight:
231.29
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing