
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate

Synonym(s):
Triphosphopyridine nucleotide sodium salt hydrate
Empirical Formula (Hill Notation):
C21H27N7NaO17P3 · xH2O
CAS Number:
Molecular Weight:
765.39 (anhydrous basis)
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Tissue
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing