
Betaine hydrochloride

Synonym(s):
(Carboxymethyl)trimethylammonium hydrochloride, Betaine hydrochloride
Linear Formula:
(CH3)3N+CH2COOH · Cl-
CAS Number:
Molecular Weight:
153.61
Products
Product Category
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3