Skip to Content
Merck
Benzyl chloromethyl ether technical, ~60% (NMR)

Benzyl chloromethyl ether

Benzyl chloromethyl ether technical, ~60% (NMR)
Synonym(s):
(Chloromethoxymethyl)benzene, Benzyloxymethyl chloride
Linear Formula:
C6H5CH2OCH2Cl
CAS Number:
Molecular Weight:
156.61

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing