
Benazepril hydrochloride

Synonym(s):
(3S)-3-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid hydrochloride, Benazepril hydrochloride
Empirical Formula (Hill Notation):
C24H28N2O5 · HCl
CAS Number:
Molecular Weight:
460.95
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing