
Atomoxetine S-isomer

Synonym(s):
(S)-N-Methyl-γ-(2-methylphenoxy)benzenepropanamine hydrochloride, (S)-Tomoxetine hydrochloride
Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing