
Aristolochia

Synonym(s):
Aristolochic acid I, TR 1736
Empirical Formula (Hill Notation):
C17H11NO7
CAS Number:
Molecular Weight:
341.27
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing