Skip to Content
Merck
Anti-SPARC antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-SPARC antibody produced in rabbit

Anti-SPARC antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-BM-40 antibody produced in rabbit, Anti-Basement-membrane protein 40 antibody produced in rabbit, Anti-ON antibody produced in rabbit, Anti-Osteonectin antibody produced in rabbit, Anti-SPARC precursor antibody produced in rabbit, Anti-Secreted protein acidic and rich in cysteine antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique