
Anti-HNK-1/N-CAM (CD57) antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-HNK-1, Anti-CD57, Anti-N-CAM
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing