![Anti-β-Tubulin antibody, Mouse monoclonal ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/163/689/6ff333e6-3327-4aa8-9dbb-e637bcdd979e/640/6ff333e6-3327-4aa8-9dbb-e637bcdd979e.jpg)
Anti-β-Tubulin antibody, Mouse monoclonal
![Anti-β-Tubulin antibody, Mouse monoclonal ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/163/689/6ff333e6-3327-4aa8-9dbb-e637bcdd979e/640/6ff333e6-3327-4aa8-9dbb-e637bcdd979e.jpg)
Synonym(s):
Anti-TUBB, Anti-Tubulin, beta, Monoclonal Anti-β-Tubulin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing