
Albendazole sulfone

Synonym(s):
Methyl [5-(propylsulfonyl)-1H-benzimidazol-2-yl]carbamate
Empirical Formula (Hill Notation):
C12H15N3O4S
CAS Number:
Molecular Weight:
297.33
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing