Skip to Content
Merck
Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate ≥93% (HPLC), powder

Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate

Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate ≥93% (HPLC), powder
Synonym(s):
β,γ-Imidoadenosine 5′-triphosphate lithium salt hydrate, AMP-PNP, ATP[β,γ-NH], Adenylyl imidodiphosphate lithium salt hydrate, App(NH)p
Empirical Formula (Hill Notation):
C10H17N6O12P3 · xLi+ · yH2O
CAS Number:
Molecular Weight:
506.20 (free acid basis)

Products

Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1