
Actein

Synonym(s):
Shengmating
Empirical Formula (Hill Notation):
C37H56O11
CAS Number:
Molecular Weight:
676.83
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing