Skip to Content
Merck
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate 96%

(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate

(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate 96%
Synonym(s):
(3-Hydroxy-3H-1,2,3-triazolo[4,5-b]pyridinato-O)tri-1-pyrrolidinyl-phosphorus hexafluorophosphate, PyAOP
Empirical Formula (Hill Notation):
C17H27F6N7OP2
CAS Number:
Molecular Weight:
521.38

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Reaction Type
Showing 1-1 of 1