
5-Hexynenitrile

Synonym(s):
5-Cyano-1-pentyne
Linear Formula:
HC≡C(CH2)3CN
CAS Number:
Molecular Weight:
93.13
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing