Skip to Content
Merck
3-Bromodiphenyl ether 97%

3-Bromodiphenyl ether

3-Bromodiphenyl ether 97%
Synonym(s):
1-Bromo-3-phenoxybenzene
Empirical Formula (Hill Notation):
C12H9BrO
CAS Number:
Molecular Weight:
249.10

Products

Product Category
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing