2-Ethyl-6-methylaniline
Synonym(s):
3-Ethyl-2-aminotoluene, 6-Ethyl-o-toluidine
Linear Formula:
C2H5C6H3(CH3)NH2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1