
17β-Estradiol-2,3,4-13C3 solution

Empirical Formula (Hill Notation):
13C3C15H24O2
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing