Merck
HomeProductsChemistry & BiochemicalsBiochemicalsChelating & Reducing Agents for Research
Chelating & Reducing Agents for Research
Structure of molecule
We offer a wide range of chelating and reducing agents in high-purity grades and diverse forms for biochemical research.
Chelators, or chelating agents, are found in biological substances and are utilized for the removal of heavy metal impurities, which can be often toxic, and restore cellular metabolism. Through their ring structures, chelators can establish two or more bonds with metal ions, resulting in the formation of water-soluble, stable complexes. Ethylenediamine, EDTA, and EGTA are well-known examples of chelators. 
Conducting both protein biochemistry analysis and oxidation-reduction reactions necessitates the use of reducers or reducing agents, also known as reductants. Reductants donate electrons in a redox chemical reaction. Several common reducing agents are DL-Dithiothreitol (DTT), sodium dithionate, DTE, TCEP, β-mercaptoethanol (BME), and Nitrilotriacetic acid (NTA).
Products
Popular Filters
Sort by Relevance
Showing 1-20 of 88
Change View
Showing 1-20 of 88
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 88 results
CHELATING AGENTS IN RESEARCH
Chelators are used as complexing agents. They are helpful in various chemical and biochemical analyses as a buffer component, nucleic acid/protein degradation inhibitor, enzymatic reaction catalyzer, affinity chromatography, and cell culture. Chelators are potent therapeutic agents to balance metabolism and associated diseases such as anemia, cancer, acute kidney disease, aging, neurodegenerative disorders, etc. In industrial, analytical, and laboratory experiments, these metal-ion chelators are used as extractants, indicators, and metal separation agents.
REDUCING AGENTS IN RESEARCH
Reducers are utilized in proteomics methods such as protein purification, denaturation, and solubilization and are particularly adept in stabilizing free sulfhydryl groups and reducing disulfide (SH) bonds in proteins and peptides. In protein purification, they act as enzyme stabilizers within buffers, preventing aggregation by cysteine residue oxidation. Reducers also serve as biological antioxidants, averting protein oxidation and denaturing ribonucleases. Research has demonstrated that using chelating reagents in combination with reducing agents supports the removal of cationic heavy metals and anionic contaminants, such as arsenic.