Merck
HomeProductsCell Culture & AnalysisSterile Lab Media Filtration & Cultureware
Sterile Lab Media Filtration & Cultureware
Critical in reducing contamination in culture, sterile laboratory media filters are used to remove particulate contamination, sterilize liquid reagents, and clarify solutions of laboratory reagents, buffers, and media. Our sterile filtration solutions feature high-performance membranes and are available in a wide variety of formats including syringe filters, vacuum-driven filter units, and bottle top filters. Stericup® filter units are available with a receiver flask for processing and storing volumes from 150 mL up to 1,000 mL.   
 A woman in a white lab coat, working with laboratory flask containing a red liquid, which is connected to another piece of equipment with blue liquid at the bottom.
Millipore® Sterile Filters
Sterile filtration devices are used to clarify and prevent microbial contamination of cell culture media, cell culture reagents, buffers, biological samples, proteinaceous samples, and more. These devices provide an alternative to autoclaving, which can degrade sensitive, heat-labile biological components. We provide small to large-volume Millipore® sterile filters for cell culture, preparation of reagents for long-term storage, and other applications.
Stericup® Vacuum Filters
Stericup® filtration devices have been the most trusted systems in media and buffer filtration for decades. The next-generation Stericup® Quick Release vacuum filtration devices present ergonomic design updates that optimize usability and streamline the filtration process while protecting essential cell cultures with the renowned performance of Millipore® membranes.   
The minds of Millipore® scientists don’t stop with the most reliable sterile filtration in the world. Our commitment to sustainability drove us to design a dependable sterile filter device that helps labs attain sustainability goals. Stericup® E and Steritop® E filters provide the same steadfast sterile filtration cell culturists worldwide have chosen for decades—but attach directly to commercial media bottles, eliminating the plastic filter funnel to reduce plastic waste up to 48%.
A collection of five laboratory volumetric tools with varying amounts of liquid. From left to right, there is a small beaker with red liquid reaching the measurement line, a graduated cylinder half-filled with red liquid, another graduated cylinder with a smaller amount of liquid at the bottom, an empty volumetric flask on top of an upside-down beaker, and finally, a large beaker half-filled with clear liquid. The tools are marked with measurement lines and are placed against a white background.
Explore More Categories
A detailed grayscale image of a cell culture plate with multiple wells, highlighting the precision and uniformity essential for cell culture experiments, part of a comprehensive collection of cell culture supplies.
Cell Culture Supplies
An extensive collection of cell culture supplies including serological pipets, cell spreaders and lifters, flasks for adherent
View Products
An assortment of Millex<sup>®</sup> syringe filters in various colors and sizes, displayed against a white background. These filters represent a range of small-volume filtration options for laboratory use. The filters come in different colors such as blue, green, yellow, purple, and red, indicating different filter types or pore sizes. Each filter has a circular shape with a conical funnel-like structure on top for attaching to a syringe. The central part of the filters is white with labeling that includes the brand name “Millex®”. Some filters have a transparent top section while others are opaque. There is also one funnel accessory included in the image which is used to assist with pouring liquids into the filter. These filters set the standard in small-volume filtration (1-200 mL), and their unsurpassed consistency in sample preparation has led to the development of many methods specifying their use.
Millex® Syringe Filters
Millex® syringe filters are available in sterile and non-sterile formats, offered in many membrane and housing options...
View Products
A laboratory setting where a researcher, wearing purple latex gloves for safety, is holding a Millicell<sup>®</sup> cell culture insert filled with a pink medium. This insert is being held above a multi-well plate, which also contains the same pink medium in its multiple wells. The focus of the image is on the cell culture insert and the multi-well plate, emphasizing their importance in cell culture applications. The background features a stainless steel surface, typical of lab environments for easy cleaning and sterilization.
Millicell® Cell Culture Inserts & Plates
Millicell®  cell culture inserts and plates, chamber slides, and multilayer cell culture flasks are...
View Products
MultiScreen® filtration plates for cell-based and biochemical assays
MultiScreen® Filter Plates
MultiScreen® filtration plates in 96-well and 384-well formats with a variety of membrane options are for high-throughput...
View Products
Shop Products
Sort by Relevance
Showing 1-20 of 52
Change View
Showing 1-20 of 52
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 52 results