Skip to Content
Merck
All Photos(1)

Key Documents

SPRFKIT7

Smoke Point Reference Fuel Blends Kit

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

Components

Kit consists of:
SPRF1 Smoke Point Reference Fuel Blend 1 for 14.7 mm (40/60 %v/v)
SPRF2 Smoke Point Reference Fuel Blend 2 for 20.2 mm (25/75 %v/v)
SPRF3 Smoke Point Reference Fuel Blend 3 for 22.7 mm (20/80 %v/v)
SPRF4 Smoke Point Reference Fuel Blend 4 for 25.8 mm (15/85 %v/v)
SPRF5 Smoke Point Reference Fuel Blend 5 for 30.2 mm (10/90 %v/v)
SPRF6 Smoke Point Reference Fuel Blend 6 for 35.4 mm (5/95 %v/v)
SPRF7 Smoke Point Reference Fuel Blend 7 for 42.8 mm (0/100 %v/v)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service