Skip to Content
Merck
All Photos(1)

Key Documents

22824

Parker® ChromGas® Zero Air Generator

Model 1000, 110 volt, output flow, 1000 cc/min

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500

feature

output flow 1000 cc/min

Looking for similar products? Visit Product Comparison Guide

General description

This ChromGas zero air generator incorporates three stages:
  • A 0.5 mm coalescing inlet filter removes particles, oil, and water
  • A heated catalyst removes hydrocarbons
  • A 0.01 mm cellulose fiber outlet filter removes residual particulate material from the product air stream
A hazard-free and economical alternative to high pressure gas cylinders, this unit should pay for itself in a year or two.

Features/Benefits
  • Eliminates need for expensive gas cylinders
  • Compact; requires less than one square foot of bench space
  • Can be used anywhere that an electrical supply is available
  • Only requires an upstream compressed air source

Maintenance
  • Clean the inlet and outlet filters every six months
  • Change the inlet and outlet filters every two years

Specifications
  • Inlet Pressure: 2-125 psig
  • Outlet Purity: <0.1 ppm hydrocarbons (as methane)
  • Outlet Pressure: 125 psig
  • Outlet Fitting: 1/8 in. compression
  • Power Needs: 110 V (60 Hz), or 230 V (50 Hz)
  • Shipping Weight (Model 1000 and 1001): 11 lb (5 Kg)
  • Shipping Weight (Model 3500 and 3501): 20 lb (9.1 Kg)
  • Dimensions (Model 1000 and 1001; H x W x D): 9.75 in. x 5.75 in. x 12 in. (25 cm x 14.7 cm x 30.8 cm)
  • Dimensions (Model 3500 and 3501; H x W x D): 12 in. x 6.75 in. x 15 in. (29.2 cm x 17.8 cm x 39.4 cm)

Marks
  • CE approved
  • UL listed
  • CSA listed
  • IEC 1010 certified

Application

Catalytic Zero air generator may be used to measure zero air count rates after field measurements, to determine atmospheric concentrations of volatile organic compound using On-line proton transfer reaction-mass spectrometry (PTR-MS). It may also be used to obtain volatile organic compound-air which could be used to dilute the standard gas to 50 ppb required to calibrate PTR-MS.

Legal Information

Chromgas is a registered trademark of Parker Hannifin
Parker is a registered trademark of Parker Hannifin

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrocarbon fluxes above a Scots pine forest canopy: measurements and modeling.
Rinne, J., et al.
Atmospheric Chemistry and Physics, 7, 3361-3372 (2007)
On-line PTR-MS measurements of atmospheric concentrations of volatile organic compounds in a European boreal forest ecosystem.
Rinne, Janne, et al.
Boreal Environment Research, 10, 425-436 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service