Skip to Content
Merck
All Photos(1)

Key Documents

SML1335

Sigma-Aldrich

Pidotimod

≥98% (HPLC)

Synonym(s):

(4R)-3-(5-Oxo-L-prolyl)-1,3-thiazolidine-4-carboxylic acid, (R)-3-((S)-5-Oxopyrrolidine-2-carbonyl)thiazolidine-4-carboxylic acid, (R)-3-[(S)-(5-Oxo-2-pyrrolidinyl) carbonyl]-thiazolidine-4-carboxylic acid, 3-L-Pyroglutamyl-L-thiaziolidine-4carboxylic acid, PGT/1A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O4S
CAS Number:
Molecular Weight:
244.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -135 to -155°, c = 1 in 6 M HCl

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

OC([C@@H]1CSCN1C([C@H]2NC(CC2)=O)=O)=O

InChI

1S/C9H12N2O4S/c12-7-2-1-5(10-7)8(13)11-4-16-3-6(11)9(14)15/h5-6H,1-4H2,(H,10,12)(H,14,15)/t5-,6-/m0/s1

InChI key

UUTKICFRNVKFRG-WDSKDSINSA-N

Biochem/physiol Actions

Pidotimod is a dipeptide immunostimulant with biological activity on both the adaptive and the innate immune responses. Pidotimod has been reported to modulate airway epithelial cells functions involved in host-virus interaction possibly through NF-kB activation, promote functional maturation of dendritic cells, and facilitate M2 macrophage polarization and function. Pidotimod has been investigated for use to increase response to recurrent respiratory infections primarily in children.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service