Skip to Content
MilliporeSigma

Skip To

RAB0502

Human Thyroid Stimulating Hormone ELISA Kit

for serum, plasma, cell culture supernatants and urine

Synonym(s):

TSH, Thyroid Stimulating Hormone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$608.00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... CGA(1081), TSHB(7252)
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

$608.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

technique(s)

ELISA: suitable, capture ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CGA(1081), TSHB(7252)

General description

The Human TSH (Thyroid Stimulating Hormone) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human TSH in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human TSH

Application

This product is for research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0196RAB0092RAB0109
Gene Information

human ... CGA(1081), TSHB(7252)

Gene Information

human ... FSTL3(10272)

Gene Information

human ... CGB(1082)

Gene Information

human ... CTLA4(1473)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 1X Assay/Sample Diluent Buffer A (Item D1)
    SDS

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters