Skip to Content
Merck
All Photos(1)

Key Documents

M6267

Sigma-Aldrich

(±)-3,4-Methylenedioxyamphetamine hydrochloride

≥98% (TLC)

Synonym(s):

(±)-α-Methyl-1,3-benzodioxole-5-ethanamine hydrochloride, MDA hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2 · HCl
CAS Number:
Molecular Weight:
215.68
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

drug control

USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: soluble

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

SMILES string

Cl[H].CC(N)Cc1ccc2OCOc2c1

InChI

1S/C10H13NO2.ClH/c1-7(11)4-8-2-3-9-10(5-8)13-6-12-9;/h2-3,5,7H,4,6,11H2,1H3;1H

InChI key

XLAOKZDOZHZWBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service