Skip to Content
Merck
All Photos(1)

Key Documents

CLS353002

Corning® Falcon® Cell Culture Dish

plate diam. × capacity 60 x 15 mm × 6 mL, Tissue Culture (TC)-treated surface, sterile

Synonym(s):

Cell Culture Dish

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.14

sterility

sterile

packaging

case of 500 ea (25 packs of 20)

manufacturer/tradename

Corning 353002

plate diam. × capacity

60 x 15 mm × 6 mL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Falcon® 60mm x 15mm Style Cell Culture Dish, Tissue Culture-Treated, Sterile, Bulk Packaged, 20/Pack, 500/Case

Application

Cell Culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service