Skip to Content
Merck
All Photos(1)

Documents

02858

Sigma-Aldrich

Ethanol

purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21

vapor density

1.59 (vs air)
1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)
44.6 mmHg

grade

absolute
purum

Assay

≥99.8% (based on denaturant-free substance)

form

liquid

autoignition temp.

683 °F

quality

absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1

expl. lim.

19 %, 60 °F
19 %

impurities

≤0.2% water

refractive index

n20/D 1.3600 (lit.)
n20/D 1.362

bp

78 °C (lit.)
78 °C

mp

-114 °C
−114 °C (lit.)

density

0.789 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

greener alternative product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D N Misra
Journal of dental research, 68(1), 42-47 (1989-01-01)
The adsorption of 4-methacryloxyethyl trimellitate anhydride (4-META) was studied from ethanol and dichloromethane onto synthetic hydroxyapatite (containing about 1.5 monolayers of physisorbed water) in order to study its role in restorative composite bonding to teeth. The adsorption isotherm of 4-META
Pierre de Thier et al.
Biochimica et biophysica acta, 1854(2), 138-145 (2014-12-17)
IgG films are widely used in the field of immunoassays, especially in (double) antibody-sandwich ELISA tests where capture antibodies are coated on surfaces like polystyrene or hydrophobic self-assembled monolayers (h-SAMs). It is critical to analyze-at a molecular scale and under
Physicochemical Properties of SnO2-Sb2 O 5 Films Prepared by the Spray Pyrolysis Technique.
Correa-Lozano B, et al.
Journal of the Electrochemical Society, 143(1), 203-209 (1996)
Spectrophotometric characteristics of chlorophylls a and b and their pheophytins in ethanol.
J F Wintermans et al.
Biochimica et biophysica acta, 109(2), 448-453 (1965-11-29)
Fabrication of thin film dye sensitized solar cells with solar to electric power conversion efficiency over 10%.
Ito S, et al.
Thin Solid Films, 516(14), 4613-4619 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service