Skip to Content
Merck
All Photos(2)

Key Documents

07846

Supelco

2,4-Dichlorophenol

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein:
742467
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

bp

209-210 °C (lit.)

mp

42-43 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccc(Cl)cc1Cl

InChI

1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H

InChI key

HFZWRUODUSTPEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

231.1 °F - closed cup

Flash Point(C)

110.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongwei Qin et al.
Analytical biochemistry, 462, 60-66 (2014-06-29)
An in situ electrochemical method was used to assess the cytotoxicity of chlorophenols using human breast cancer (MCF-7) and cervical carcinoma (HeLa) cells as models. On treatment with different chlorophenols, the electrochemical responses of the selected cells, resulting from the
Mir Mahdi Abolghasemi et al.
Journal of separation science, 37(23), 3526-3532 (2014-09-13)
A fiber-coated polypyrrole-montmorillonite nanocomposite was prepared for solid-phase microextraction. The fiber coating can be prepared easily; it is mechanically stable and exhibits relatively high thermal stability. The prepared fiber was evaluated for the extraction of some phenolic compounds from aqueous
Dandan Ge et al.
Talanta, 132, 132-136 (2014-12-06)
An ultra-hydrophobic ionic liquid, 1-hexyl-3-methylimidazolium tris(pentafluoroethyl)trifluorophosphate ([HMIM][FAP]) was immobilized in the pores of a polypropylene hollow fiber for liquid-liquid-liquid microextraction (HF-LLLME) of chlorophenols (CPs) (4-chloro-3-methylphenol, 2,4-dichlorophenol and 2,4,6-trichlorophenol). The analytes were first extracted from 10 ml of water sample into
Xijuan Chen et al.
The Science of the total environment, 505, 39-46 (2014-10-13)
Aerobic degradation experiments of Triclosan were performed in activated sludge to identify possible transformation products for this compound. During 7 days, the formation of biotransformation products such as 2,4-Dichlorophenol, 4-Chlorocatechol, 5-Hydroxy-Triclosan and other Monohydroxy-Triclosan derivatives as well as Dihydroxy-Triclosan-derivatives were
Chen Fan et al.
Food chemistry, 174, 446-451 (2014-12-23)
In-situ ionic liquid-dispersive liquid-liquid microextraction (IL-DLLME) method was developed as a pretreatment method for the detection of six chlorophenols (CPs) in honey samples. The hydrophobic ionic liquid [C4MIM][NTf2], formed in-situ by the hydrophilic ionic liquid [C4MIM][BF4] and the ion exchange

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service