Skip to Content
Merck
All Photos(1)

Key Documents

AB9900

Sigma-Aldrich

Anti-IDO Antibody

Chemicon®, from rabbit

Synonym(s):

Indoleamine 2,3-dioxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Ido1(15930)

Specificity

IDO [Indoleamine 2,3-dioxygenase], mouse.

Immunogen

Recombinant mouse IDO.

Application

Anti-IDO Antibody detects level of IDO & has been published & validated for use in ELISA & WB.
Western blot: 1:2,000-1:5,000 using ECL. Reacts with the ~45 kDa IDO protein. Suggested blocking and dilution buffer is PBST with 0.05% Tween 20 with 5% skim milk. Suggested incubation time is 1 hour at room temperature.

ELISA, direct and indirect: 1:2,000-1:10,000.

Optimal working dilutions must be determined by the end user.

Target description

45 kDa

Linkage

Replaces: 04-1056

Physical form

Format: Purified
Liquid on PBS, pH 7.4.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph-Martin Ufermann et al.
Frontiers in cellular and infection microbiology, 9, 184-184 (2019-06-25)
Toxoplasma gondii (T. gondii) is an obligate intracellular parasite and belongs to the phylum Apicomplexa. T. gondii is of medical and veterinary importance, because T. gondii causes the parasitic disease toxoplasmosis. In human cells, the interferon-gamma inducible indoleamine 2,3-dioxygenase 1
Activation of the alternative NF?B pathway improves disease symptoms in a model of Sjogren's syndrome.
Gilboa-Geffen, A; Wolf, Y; Hanin, G; Melamed-Book, N; Pick, M; Bennett, ER; Greenberg et al.
Testing null
Giorgia Renga et al.
Nature communications, 14(1), 5753-5753 (2023-09-17)
The aromatic amino acid L-tryptophan (Trp) is essentially metabolized along the host and microbial pathways. While much is known about the role played by downstream metabolites of each pathways in intestinal homeostasis, their role in lung immune homeostasis is underappreciated.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service