Skip to Content
Merck
All Photos(1)

Key Documents

958650

Supelco

Spiral gas feeding tube (Nylon)

L 180 cm, fluoroelastomer FFKM seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.51

material

fluoroelastomer FFKM seal

L

180 cm

shipped in

ambient

storage temp.

room temp

Related Categories

General description

To meet our customers’ individual installation requirements resulting from their specific operating environments, we offer tubings with different endings for various applications. The tubing range also includes spiral tubings for gas supply (8x6mm) that are equipped with a coupling (female) Rectus Type 21 and an open end. This gas feeding tube has a working length of 180 cm and can be directly connected to the threaded adapters 9.67100.9002 and 9.67100.9003.
with rapid action-connector, FFKM sealing, length: 180 cm

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service