194437
ABTS™ Chromophore, Diammonium Salt
Synonym(s):
ABTS™ Chromophore, Diammonium Salt
About This Item
Recommended Products
form
solid
Quality Level
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
solubility
0.1 M sodium citrate, pH 4.2: 1 mg/mL
shipped in
ambient
storage temp.
2-8°C
InChI
1S/C18H18N4O6S4.2H3N/c1-3-21-13-7-5-11(31(23,24)25)9-15(13)29-17(21)19-20-18-22(4-2)14-8-6-12(32(26,27)28)10-16(14)30-18;;/h5-10H,3-4H2,1-2H3,(H,23,24,25)(H,26,27,28);2*1H3/b19-17-,20-18-;;
InChI key
OHDRQQURAXLVGJ-AXMZSLBLSA-N
General description
Warning
Reconstitution
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service