Skip to Content
Merck
All Photos(1)

Key Documents

1.14976

Supelco

Chlorine Test Kit, colorimetric

0.25-15 mg/L (Cl₂), MQuant®

Synonym(s):

Chlorine Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Chlorine Test, colorimetric, 0.25-15 mg/L (Cl2), for use with MCOLORTEST®

product line

MColortest®

Quality Level

specific analyte(s)

chlorine

measuring range

0.25-15 mg/L (Cl2)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

(free chlorine) Method: colorimetric, DPD, with color-disk comparator 0.25 - 0.50 - 0.75 - 1 - 2 - 4 - 8 - 10 - 15 mg/l Cl MQuant®
Chlorine containing disinfectants are most widely used. They are applied particularly to disinfect drinking water and swimming pool water. The MQuant® Chlorine test (formally MColortest®) can be used for the determination of free and total chlorine in
  • Groundwater and surface water
  • Drinking water and mineral water
  • Waters from aquaculture
  • Wastewater
  • Electroplating wastewater
  • Disinfectant solutions

This test is not suited for seawater.

Method
In weakly acidic solution free chlorine reacts with dipropyl-p-phenylenediamine (DPD) to form a red-violet dye. In the presence of potassium iodide, also combined chlorine is measured in this reaction. The chlorine concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.
Using the refill pack 1.14977 the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Application


  • 3,3′,5,5′-tetramethylbenzidine as a multi-colorimetric indicator of chlorine in water in line with health guideline values: This study focuses on using 3,3′,5,5′-tetramethylbenzidine as an indicator for the presence of chlorine in water, providing a method to ensure compliance with health guidelines through a visual and straightforward test (Palladino et al., 2020).

  • Analysis of free chlorine in aqueous solution at very low concentration with lateral flow tests: This paper introduces a method for testing free chlorine concentrations in water at very low levels using lateral flow assays, which are crucial for monitoring water quality and ensuring safety standards (Schwenke et al., 2019).

  • Some factors influencing the stability of Sterilox, a super-oxidised water: This article, while focused on the stability of Sterilox as a disinfectant, deals with the broader context of using chlorine solutions in settings requiring stringent sanitation measures, highlighting the importance of regular chlorine testing to ensure efficacy (Rossi-Fedele et al., 2011).


Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

AQA Standard for free chlorine - Preparation of a standard solution for free chlorine

Preparation of a standard solution for free Chlorine according to DIN EN ISO 7393

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service